Please login to your online account to display your discounted pricing

Sino Biological™ CA2 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

$96.25 - $315.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias CA-II, CAII, Car2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10478H08E25

DFS Item
 
life technologies
LS10478H08E25
5 x 5μg Each for $206.00

10478H08E5

DFS Item
 
life technologies
LS10478H08E5
5μg Each for $96.25

10478H08E50

DFS Item
 
life technologies
LS10478H08E50
50μg Each for $315.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias CA-II, CAII, Car2

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.