Please login to your online account to display your discounted pricing

Sino Biological™ CA2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

Manufacturer: life technologies  LS10478H08E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10478H08E50

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 30kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Gene Alias CA-II, CAII, Car2
Common Name Carbonic anhydrase II
Gene Symbol CA2
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.