Please login to your online account to display your discounted pricing

Sino Biological™ CA2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human CA2 (NP_000058.1) (Met 1-Lys 260) fused to the polyhistidinetag at the C-terminus

Manufacturer: life technologies  LS10478H08E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10478H08E5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Validated Application Assay Standard
Protein Subtype Other Proteins
Protein Tag His Tag
Purity ≥90% by SDS-PAGE
Common Name Carbonic anhydrase II
Expression System E. coli
Format Lyophilized
Gene Alias CA-II, CAII, Car2
Protein Family Other Proteins
Molecular Weight 30kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol CA2
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Carbonic anhydrase II (CA2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its esterase activity, the specific activity is >500pmoles/min./μg
  • Formulation: Lyophilized in 20mM Tris, 0.5M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.