missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BTC Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human BTC (P35070-1) (Asp 32-Tyr 111) fused to a polyhistide tag at the C-terminus

$502.00 - $502.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias BTC
Gene ID (Entrez) 685
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12192H08E50 Life Technologies
LS12192H08E50
50μg Each for $502.00
Specifications

Description

Betacellulin (BTC), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
Lyophilized
685
PAGE
BTC