Please login to your online account to display your discounted pricing

Sino Biological™ BST1 (CD157) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human BST1 (NP_004325.2), Met 1-Lys 292, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10060H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10060H08H25

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Molecular Weight 31.4kDa
Quantity 5 x 5μg
Protein Family CD Proteins
Protein Subtype Glycoproteins
Purity ≥90% by SDS-PAGE
Gene Alias CD157, BST1
Common Name Bone marrow stromal cell antigen 1
Protein Tag His Tag
Validated Application Assay Standard
Gene ID (Entrez) 683
Expression System Mammalian
Format Lyophilized
Structural Form Truncated/Recombinant/Cell Surface Protein
Endotoxin Concentration <1 EU/μg
Gene Symbol BST1
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

Bone marrow stromal cell antigen 1 (BST1/CD157), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.