missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BSG (CD147) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 209) of mouse BSG isoform 2 (NP_001070652.1) fused to a polyhistidine tag at the C-terminus

$256.00 - $1,246.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI115436, AI325119, CD147, EMMPRIN, HT-7, Bsg
Gene ID (Entrez) 12215
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50332M08H25 View Documents Invitrogen
LS50332M08H25
5 x 5μg Each for $256.00
 
50332M08HL View Documents Invitrogen
LS50332M08H250
5 x 50μg Each for $1,246.00
 
Description

Description

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
12215
<1 EU/μg
AI115436, AI325119, CD147, EMMPRIN, HT-7, Bsg

For Research Use Only. Not for use in diagnostic procedures.