Please login to your online account to display your discounted pricing

Sino Biological™ BSG (CD147) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 205) of human BSG precursor (NP_940991.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10186H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10186H08H50

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 682
Molecular Weight 21.6kDa
Purity ≥90% by SDS-PAGE
Quantity 50μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Glycoproteins
Endotoxin Concentration <1 EU/μg
Gene Alias BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN
Common Name Basigin
Gene Symbol BSG
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
Protein Tag His Tag

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature.