missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BSG (CD147) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 205) of human BSG precursor (NP_940991.1) fused to the Fc region of human IgG1 at the C-terminus

$291.00 - $291.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN
Gene ID (Entrez) 682
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10186H02H25 View Documents Invitrogen
LS10186H02H25
5 x 5μg Each for $291.00
 
Description

Description

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
682
<1 EU/μg
BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN

For Research Use Only. Not for use in diagnostic procedures.