Please login to your online account to display your discounted pricing

Sino Biological™ BSG (CD147) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 205) of human BSG precursor (NP_940991.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10186H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10186H02H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 46.8kDa
Purity ≥90% by SDS-PAGE
Gene Symbol BSG
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Validated Application Assay Standard
Quantity 5μg
Gene ID (Entrez) 682
Common Name Basigin
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Protein Subtype Glycoproteins
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Gene Alias BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN
Protein Tag IgG1 Fc Tag
Protein Family CD Proteins

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.