missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BSG (CD147) Recombinant Human Protein, DDDDK Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 205) of human BSG (NP_940991.1) fused to a DDDDK tag at the C-terminus

$137.00 - $1,704.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN
Gene ID (Entrez) 682
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10186HCCH25 Life Technologies
LS10186HCCH25
5 x 5μg Each for $338.00
10186HCCH5 Life Technologies
LS10186HCCH5
5μg Each for $137.00
10186HCCH50 Life Technologies
LS10186HCCH50
50μg Each for $540.00
10186HCCHL Life Technologies
LS10186HCCH250
5 x 50μg Each for $1,704.00
Specifications

Description

Basigin (BSG/CD147), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
682
<1 EU/μg
BSG, M6, OK, 5F7, TCSF, CD147, EMMPRIN