missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BMPR2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 151) of human BMPR2 (NP_001195.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$78.75 - $906.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BMPR2, BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK
Gene ID (Entrez) 659
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10551H03H25 Life Technologies
LS10551H03H25
5 x 5μg Each for $208.00
10551H03H5 Life Technologies
LS10551H03H5
5μg Each for $78.75
10551H03H50 Life Technologies
LS10551H03H50
50μg Each for $226.00
10551H03HL Life Technologies
LS10551H03H250
5 x 50μg Each for $906.00
Specifications

Description

Bone morphogenetic protein receptor type II (BMPR2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
659
<1 EU/μg
BMPR2, BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK