Please login to your online account to display your discounted pricing

Sino Biological™ BMPR2 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ile 151) of human BMPR2 (NP_001195.2) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10551H03H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10551H03HL

  • / Each
 

Description & Specifications

Specifications

Expression System Mammalian
Format Lyophilized
Structural Form Truncated/Recombinant
Protein Tag His Tag/IgG1 Fc Tag
Protein Family Growth Factors & Receptors
Purity ≥85% by SDS-PAGE
Gene ID (Entrez) 659
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Protein Subtype Serine/Threonine Kinases|Growth Factor Receptors
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Gene Symbol BMPR2
Quantity 5 x 50μg
Common Name Bone morphogenetic protein receptor, type II (serine/threonine kinase)
Gene Alias BMPR2, BMPR-II, BMPR3, BMR2, BRK-3, FLJ41585, FLJ76945, PPH1, T-ALK
Molecular Weight 42kDa

Bone morphogenetic protein receptor type II (BMPR2), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.