missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BMPR1A Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 152) of mouse ALK3 (NP_033888.2) precursor fused to a polyhistidine tag at the C-terminus

$209.00 - $209.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias Bmpr1a, 1110037I22Rik, ALK3, AU045487, BMPR-IA, Bmpr
Gene ID (Entrez) 12166
View More Specs

Includes: Five vials of protein in suspension buffer

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50078M08H25 View Documents Invitrogen
LS50078M08H25
5 x 5μg Each for $209.00
 
Description

Description

Bone Morphogenetic Protein Receptor, Type Ia (BMPR1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit recombinant human BMP4 induced activity in MC3T3-E1 Mouse osteoblastic cells; the ED50 for this effect is typically 0.5-2μg/mL in the presence of 50ng/mL of recombinant human BMP4
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
12166
PAGE
Bmpr1a, 1110037I22Rik, ALK3, AU045487, BMPR-IA, Bmpr

For Research Use Only. Not for use in diagnostic procedures.