Please login to your online account to display your discounted pricing

Sino Biological™ BMPR1A, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 152) of human ALK3 (NP_004320.2) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$314.00 - $314.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias ACVRLK3, ALK3, CD292, BMPR1A
Gene ID (Entrez) 657
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10446H03H50

DFS Item
 
life technologies
LS10446H03H50
50μg Each for $314.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Gene Alias ACVRLK3, ALK3, CD292, BMPR1A
Gene ID (Entrez) 657

Bone Morphogenetic Protein Receptor, Type IA (BMPR1A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its ability to inhibit BMP4 induced activity in MC3T3-E1 Mouse osteoblastic cells, the ED50 for this effect is typically 0.1-0.3μg/mL in the presence of 50ng/mL of recombinant human BMP4
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.