missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BMP5 Recombinant Human Protein, N-hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the carboxy-terminal domain (Gln 324-His 454) (the mature chain) of human BMP5 (NP_066551.1) fused to Fc region of human IgG1 at the N-terminus

$1,126.00 - $1,126.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BMP5, mgC34244
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10214H01H50 Life Technologies
LS10214H01H50
50μg Each for $1,126.00
Specifications

Description

Bone Morphogenetic Proteins 5 (BMP5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
BMP5, mgC34244
PAGE
Lyophilized
653