Please login to your online account to display your discounted pricing

Sino Biological™ BLMH Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human BLMH (NP_000377.1) (Ser 2-Glu 455) fused to a polyhistidinetag at the N-terminus

$238.00 - $4,235.00

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias NME1, AWD, GAAD, NB, NBS, NDPK-A, NDPKA, nM23, nM23-H1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11333H07E25

DFS Item
 
life technologies
LS11333H07E25
5 x 5μg Each for $708.00

11333H07E50

DFS Item
 
life technologies
LS11333H07E50
50μg Each for $1,182.00

11333H07E5

DFS Item
 
life technologies
LS11333H07E5
5μg Each for $238.00

11333H07EL

DFS Item
 
life technologies
LS11333H07E250
5 x 50μg Each for $4,235.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Format Lyophilized
Gene Alias NME1, AWD, GAAD, NB, NBS, NDPK-A, NDPKA, nM23, nM23-H1

Bleomycin hydrolase (BLMH), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications., or in other research applications.

  • Activity: Measured by its ability to hydrolyze Met-AMC; the specific activity is >500pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.