missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BCL2L2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus

$211.00 - $211.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
Gene ID (Entrez) 599
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10059H08E25 View Documents Invitrogen
LS10059H08E25
5 x 5μg Each for $211.00
 
Description

Description

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 25mM HEPES, 100mM KCl, 10% Glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
599
PAGE
BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271

For Research Use Only. Not for use in diagnostic procedures.