Please login to your online account to display your discounted pricing

Sino Biological™ BCL2L2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10059H08E50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10059H08E50

  • / Each
 

Description & Specifications

Specifications

Quantity 50μg
Storage Requirements Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene ID (Entrez) 599
Purification Method PAGE
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Expression System E. coli
Protein Tag His Tag
Gene Alias BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
Purity ≥90% by SDS-PAGE
Format Frozen
Common Name Bcl2-Like 2
Structural Form Full Length/Recombinant
Molecular Weight 20kDa
Gene Symbol BCL2L2
Protein Family Other Proteins

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: 25mM HEPES, 100mM KCl, 10% Glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.