missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BCL2L2 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10059H08E25

 View more versions of this product

Catalog No. 10059H08E25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; BCL2L2 Recombinant Human Protein, His tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 25mM HEPES, 100mM KCl, 10% Glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
599
PAGE
5 x 5μg
Full Length/Recombinant
BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
BCL2L2
Other Proteins
His Tag
Frozen
20kDa
≥90% by SDS-PAGE
Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
Bcl2-Like 2
Assay Standard
E. coli