missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BACE1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed as a secreted protein from a DNA sequence encoding the extracellular domain (Met 1-Thr 457) of human BACE1 (NP_036236.1) fused to the Fc region of human IgG1 at the C-terminus

$233.00 - $233.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias BACE1, ASP2, BACE, HSPC104, FLJ90568, KIAA1149
Gene ID (Entrez) 23621
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10064H02H25 Life Technologies
LS10064H02H25
5 x 5μg Each for $233.00
Specifications

Description

Beta-secretase 1, recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing enzymatic assays, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, Mca-SEVNLDAEFRK(Dpn)RR-NH2; the specific activity is >0.5 pmoles/min./μg
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Lyophilized
23621
<1 EU/μg
BACE1, ASP2, BACE, HSPC104, FLJ90568, KIAA1149