missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ BACE1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed as a secreted protein from a DNA sequence encoding the extracellular domain (Met 1-Thr 457) of human BACE1 (NP_036236.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Life Technologies LS10064H02H25

 View more versions of this product

Catalog No. 10064H02H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; BACE1 Recombinant Human Protein, hIgG1-Fc tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 85% by SDS-PAGE

Beta-secretase 1, recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing enzymatic assays, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, Mca-SEVNLDAEFRK(Dpn)RR-NH2; the specific activity is >0.5 pmoles/min./μg
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
23621
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Proteases
BACE1, ASP2, BACE, HSPC104, FLJ90568, KIAA1149
BACE1
Other Proteins
IgG1 Fc Tag
Lyophilized
75kDa
5 x 5μg
Truncated/Recombinant
<1 EU/μg
beta-site APP-cleaving enzyme 1
Assay Standard
Mammalian