missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ B2M Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse B2M (P01887) (Met 1-Met 119), without the propeptide, fused to a C-terminal polyhistidine tag

$144.00 - $358.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-34E24.5, Ly-m11, beta2-m, beta2m
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50957M08H25 Life Technologies
LS50957M08H25
5 x 5μg Each for $358.00
50957M08H5 Life Technologies
LS50957M08H5
5μg Each for $144.00
Specifications

Description

Beta-2-microglobulin (B2M), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
RP23-34E24.5, Ly-m11, beta2-m, beta2m
PAGE
Lyophilized
12010