missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

B2M Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human B2M (NP_004039.1) (Met 1-Met 119) fused to a polyhistidine tag at the C-terminus

$179.00 - $481.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CDABP0092
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11976H08H50 View Documents Invitrogen
LS11976H08H50
50μg Each for $179.00
 
11976H08HL View Documents Invitrogen
LS11976H08H250
5 x 50μg Each for $481.00
 
Description

Description

Beta-2-microglobulin (B2M), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
CDABP0092
PAGE
Lyophilized
567

For Research Use Only. Not for use in diagnostic procedures.