missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

B2M Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human B2M (NP_004039.1) (Met 1-Met 119) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen LS11976H08H250

 View more versions of this product

Catalog No. 11976H08HL



Description

Description

Beta-2-microglobulin (B2M), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
13.5kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
CDABP0092
B2M
MHC (Major Histocompatibility Complex) Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
567
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
Beta-2-microglobulin
His Tag
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.