Please login to your online account to display your discounted pricing

Sino Biological™ ARSA Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 506) of mouse ARSA precursor (NP_033843.2) fused to the polyhistidine tag at the C-terminus

$159.00 - $2,530.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Arsa, AS-A, ASA, AW212749, As-2, As2, C230037L18Rik, TISP73
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50018M08H25

DFS Item
 
life technologies
LS50018M08H25
5 x 5μg Each for $475.00

50018M08H50

DFS Item
 
life technologies
LS50018M08H50
50μg Each for $720.00

50018M08H5

DFS Item
 
life technologies
LS50018M08H5
5μg Each for $159.00

50018M08HL

DFS Item
 
life technologies
LS50018M08H250
5 x 50μg Each for $2,530.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Arsa, AS-A, ASA, AW212749, As-2, As2, C230037L18Rik, TISP73

Arylsulfatase A (ARSA), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave p-Nitrocatechol Sulfate (PNCS); the specific activity is >100pmoles/min./μg
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.