Please login to your online account to display your discounted pricing

Sino Biological™ ARG1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ARG1 (P05089-1) (Met 1-Lys 322) fused to a polyhistidine tag at the C-terminus

$716.00 - $716.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias ARG1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11558H08H25

DFS Item
 
life technologies
LS11558H08H25
5 x 5μg Each for $716.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Frozen
Gene Alias ARG1

Arginase Liver (ARG1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by the production of urea during the hydrolysis of arginine; the specific activity is >35,000pmole/min./μg
  • Formulation: 20mM Tris, 150mM NaCl, 0.2μg/μL BSA, 20% glycerol, 1mM DTT, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.