Please login to your online account to display your discounted pricing

Sino Biological™ ARG1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human ARG1 (P05089-1) (Met 1-Lys 322) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11558H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11558H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 36.2kDa
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias ARG1
Common Name Arginase, liver
Gene Symbol ARG1
Protein Tag His Tag
Validated Application Assay Standard
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen

Arginase Liver (ARG1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by the production of urea during the hydrolysis of arginine; the specific activity is >35,000pmole/min./μg
  • Formulation: 20mM Tris, 150mM NaCl, 0.2μg/μL BSA, 20% glycerol, 1mM DTT, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.