missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

APOE Recombinant Human Protein, N-TRX.His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human APOE (P02649) (Lys 19-His 317) fused to a Trx and a polyhistidine tag at the N-terminus

$90.00 - $192.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias AD2, LDLCQ5, LPG, mgC1571
Gene ID (Entrez) 348
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10817H30E25 View Documents Invitrogen
LS10817H30E25
5 x 5μg Each for $90.00
 
10817H30EL View Documents Invitrogen
LS10817H30E250
5 x 50μg Each for $192.00
 
Description

Description

Apolipoprotein E (APOE), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: PBS, 40% Glycerol, 0.5mM TCEP, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
348
PAGE
AD2, LDLCQ5, LPG, mgC1571

For Research Use Only. Not for use in diagnostic procedures.