Please login to your online account to display your discounted pricing

Sino Biological™ APOE Recombinant Human Protein, N-TRX.His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human APOE (P02649) (Lys 19-His 317) fused to a Trx and a polyhistidine tag at the N-terminus

Manufacturer: life technologies  LS10817H30E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10817H30E5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 51.3kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Full Length/Recombinant
Protein Subtype Other Proteins
Gene Alias AD2, LDLCQ5, LPG, mgC1571
Common Name Apolipoprotein E
Gene Symbol APOE
Protein Tag Thioredoxin/His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Frozen
Gene ID (Entrez) 348

Apolipoprotein E (APOE), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: PBS, 40% Glycerol, 0.5mM TCEP, pH 7.4
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.