missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

APOA1 Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse APOA1 (Q00623) (Met 1-Gln 264) fused to the Fc region of human IgG1 at the C-terminus

$195.00 - $195.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Alp-1, Apoa-1, Brp-14, Ltw-1, Lvtw-1, mgC102525, Sep-1, Sep-2, Sep2
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50918M02H25 View Documents Invitrogen
LS50918M02H25
5 x 5μg Each for $195.00
 
Description

Description

Apolipoprotein A-I (APOA1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse APOAI at 10μg/mL (100μL/well) can bind biotinylated human SCARB1; the EC50 of biotinylated human SCARB1 is 0.27μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
Lyophilized
Alp-1, Apoa-1, Brp-14, Ltw-1, Lvtw-1, mgC102525, Sep-1, Sep-2, Sep2

For Research Use Only. Not for use in diagnostic procedures.