missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

APOA1 (APOAI) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro form of human APOA1 (CAA26097.1) (Met 1-Gln 267) was fused with Fc region of human IgG1 at the C-terminus

$196.00 - $196.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias APOA1, mgC117399
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10686H02H50 View Documents Invitrogen
LS10686H02H50
50μg Each for $196.00
 
Description

Description

Apolipoprotein A-I (APOAI), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to inhibit proliferation of HeLa human cervical epithelial carcinoma cells, the ED50 for this effect is typically 2.5-10μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
APOA1, mgC117399

For Research Use Only. Not for use in diagnostic procedures.