Please login to your online account to display your discounted pricing

Sino Biological™ APOA1 (APOAI) Recombinant Human Protein, hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the pro form of human APOA1 (CAA26097.1) (Met 1-Gln 267) was fused with Fc region of human IgG1 at the C-terminus

Manufacturer: life technologies  LS10686H02H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10686H02H5

  • / Each
 

Description & Specifications

Specifications

Gene Alias APOA1, mgC117399
Format Lyophilized
Molecular Weight 55kDa
Protein Family Other Proteins
Purity ≥90% by SDS-PAGE
Protein Subtype Other Proteins
Validated Application Assay Standard
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Quantity 5μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Expression System Mammalian
Gene Symbol APOA1
Endotoxin Concentration <1 EU/μg
Common Name Apolipoprotein A-I
Protein Tag IgG1 Fc Tag

Apolipoprotein A-I (APOAI), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit proliferation of HeLa human cervical epithelial carcinoma cells, the ED50 for this effect is typically 2.5-10μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.