Please login to your online account to display your discounted pricing

Sino Biological™ ANXA5 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human ANXA5 (P08758) (Met 1-Asp 320)

Manufacturer: life technologies  LS10448HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10448HNAE25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 35.9kDa
Purification Method PAGE
Purity ≥85% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Structural Form Full Length/Recombinant
Protein Subtype Phospholipases
Endotoxin Concentration <1 EU/μg
Gene Alias ANX5, ENX2, PP4
Common Name Annexin A5
Gene Symbol ANXA5
Validated Application Assay Standard
Protein Family Other Proteins
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 308

Annexin A5 (ANXA5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 240mM NaCl, pH 8.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.