missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ANPEP Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the metalloprotease domain (Lys 69-Lys 967, 603 Met/Ile) of the mature form of human ANPEP (NP_001141.2) with the signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

$716.00 - $1,230.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ANPEP, APN, CD13, LAP1, PEPN, gp150
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10051H08H25 View Documents Invitrogen
LS10051H08H25
5 x 5μg Each for $716.00
 
10051H08H50 View Documents Invitrogen
LS10051H08H50
50μg Each for $1,230.00
 
Description

Description

Alanyl Amonipeptidase (ANPEP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Ala-7-amido-4-methylcoumarin (Ala-AMC); the specific activity is >2500 pmoles/min./μg
  • Formulation: Lyophilized in 25mM MES, 0.15M NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
ANPEP, APN, CD13, LAP1, PEPN, gp150

For Research Use Only. Not for use in diagnostic procedures.