missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ANGPTL5 Recombinant Human Protein, GST Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 26-Lys 388) of human ANGPTL5 (NP_835228.2) fused to a GST tag at the N-terminus

$359.00 - $359.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ANGPTL5
Gene ID (Entrez) 253935
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11094H09B25 View Documents Invitrogen
LS11094H09B25
5 x 5μg Each for $359.00
 
Description

Description

Angiopoietin-like Protein 5 (ANGPTL5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 0.5mM PMSF, 1mM reduced Glutathione, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
253935
<1 EU/μg
ANGPTL5

For Research Use Only. Not for use in diagnostic procedures.