Please login to your online account to display your discounted pricing

Sino Biological™ ANGPTL5 Recombinant Human Protein, GST tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 26-Lys 388) of human ANGPTL5 (NP_835228.2) fused to a GST tag at the N-terminus

Manufacturer: life technologies  LS11094H09B5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11094H09B5

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 253935
Validated Application Assay Standard
Molecular Weight 68kDa
Structural Form Recombinant
Gene Alias ANGPTL5
Protein Subtype Growth Factor Receptors
Endotoxin Concentration <1 EU/μg
Protein Tag GST Tag
Common Name Angiopoietin-like 5
Protein Family Growth Factors & Receptors
Expression System Insect
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Symbol ANGPTL5
Purity ≥90% by SDS-PAGE
Quantity 5μg

Angiopoietin-like Protein 5 (ANGPTL5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 0.5mM PMSF, 1mM reduced Glutathione, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.