missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ANGPTL5 Recombinant Human Protein, GST tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 26-Lys 388) of human ANGPTL5 (NP_835228.2) fused to a GST tag at the N-terminus

Manufacturer:  Life Technologies LS11094H09B25

 View more versions of this product

Catalog No. 11094H09B25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; ANGPTL5 Recombinant Human Protein, GST tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Angiopoietin-like Protein 5 (ANGPTL5), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 0.5mM PMSF, 1mM reduced Glutathione, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
253935
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Growth Factor Receptors
ANGPTL5
ANGPTL5
Growth Factors & Receptors
GST Tag
Lyophilized
68kDa
5 x 5μg
Recombinant
<1 EU/μg
Angiopoietin-like 5
Assay Standard
Insect