Please login to your online account to display your discounted pricing

Sino Biological™ AMICA1 (JAML) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AMICA1 (NP_001091996.1) (Met 1-Leu 275) fused to a polyhistidine tag at the C-terminus

$97.00 - $97.00

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias JAML, AMICA, Gm638, CREA7-1, CREA7-4, FLJ37080, mgC118814, MGC118815, AMICA1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10120H08H5

DFS Item
 
life technologies
LS10120H08H5
5μg Each for $97.00
Description & Specifications

Specifications

Purity ≥85% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias JAML, AMICA, Gm638, CREA7-1, CREA7-4, FLJ37080, mgC118814, MGC118815, AMICA1

Adhesion molecule, interacts with CXADR antigen 1 (AMICA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit NGF-induced proliferation of TF1 human erythroleukemic cells
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.