Please login to your online account to display your discounted pricing

Sino Biological™ AMICA1 (JAML) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human AMICA1 (NP_001091996.1) (Met 1-Leu 275) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10120H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10120H08H5

  • / Each
 

Description & Specifications

Specifications

Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Molecular Weight 30.5kDa
Purity ≥85% by SDS-PAGE
Protein Subtype Intercellular Adhesion Molecule
Endotoxin Concentration <1 EU/μg
Gene Alias JAML, AMICA, Gm638, CREA7-1, CREA7-4, FLJ37080, mgC118814, MGC118815, AMICA1
Common Name Adhesion molecule, interacts with CXADR antigen 1
Gene Symbol AMICA1
Protein Tag His Tag
Validated Application Assay Standard

Adhesion molecule, interacts with CXADR antigen 1 (AMICA1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to inhibit NGF-induced proliferation of TF1 human erythroleukemic cells
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.