missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALPL Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human tissue-nonspecific alkaline phosphatase (NP_000469.3) (Met 1-Ser 502) fused to a C-terminal polyhistidine tag

$632.00 - $632.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ALPL, AP-TNAP, FLJ40094, FLJ93059, HOPS, mgC161443, mgC167935, TNAP, TNSALP
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10440H08H25 View Documents Invitrogen
LS10440H08H25
5 x 5μg Each for $632.00
 
Description

Description

Alkaline Phosphatase, liver/bone/kidney (ALPL), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 25mM Tris, 0.15M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
ALPL, AP-TNAP, FLJ40094, FLJ93059, HOPS, mgC161443, mgC167935, TNAP, TNSALP
PAGE
Lyophilized
249

For Research Use Only. Not for use in diagnostic procedures.