Please login to your online account to display your discounted pricing

Sino Biological™ ALPL Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human tissue-nonspecific alkaline phosphatase (NP_000469.3) (Met 1-Ser 502) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10440H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10440H08H5

  • / Each
 

Description & Specifications

Specifications

Purification Method PAGE
Structural Form Full Length/Recombinant
Endotoxin Concentration <1 EU/μg
Gene Symbol ALPL
Termination C-Terminal
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 55kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Format Lyophilized
Validated Application Assay Standard
Expression System Mammalian
Protein Family Other Proteins
Protein Tag His Tag
Quantity 5μg
Gene ID (Entrez) 249
Gene Alias ALPL, AP-TNAP, FLJ40094, FLJ93059, HOPS, mgC161443, mgC167935, TNAP, TNSALP
Common Name Alkaline Phosphatase, liver/bone/kidney

Alkaline Phosphatase, liver/bone/kidney (ALPL), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: 25mM Tris, 0.15M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.