missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALDH7A1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ALDH7A1 isoform 2 (P49419-2) (Ser 2-Gln 511) fused to a polyhistidine tag at the N-terminus

$152.00 - $152.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias ATQ1, EPD, FLJ11738, FLJ92814, PDE, ALDH7A1
Gene ID (Entrez) 501
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11614H07E5 View Documents Invitrogen
LS11614H07E5
5μg Each for $152.00
 
Description

Description

Aldehyde Dehydrogenase 7 Family, Member A1 (ALDH7A1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 500mM NaCl, 20% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Frozen
501
PAGE
ATQ1, EPD, FLJ11738, FLJ92814, PDE, ALDH7A1

For Research Use Only. Not for use in diagnostic procedures.