missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALDH7A1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ALDH7A1 isoform 2 (P49419-2) (Ser 2-Gln 511) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Invitrogen LS11614H07E5

 View more versions of this product

Catalog No. 11614H07E5



Description

Description

Aldehyde Dehydrogenase 7 Family, Member A1 (ALDH7A1), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 50mM Tris, 500mM NaCl, 20% glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Frozen
56kDa
≥90% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
ATQ1, EPD, FLJ11738, FLJ92814, PDE, ALDH7A1
ALDH7A1
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
501
PAGE
5μg
Full Length/Recombinant
Aldehyde Dehydrogenase 7 Family, Member A1
His Tag
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.