missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALDH1A3 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ALDH1A3 (NP_000684.2) (Ala 2-Pro 512) fused to a polyhistide tag at the N-terminus

$654.00 - $654.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias ALDH1A6, ALDH6, RALDH3
Gene ID (Entrez) 220
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11636H07E50 View Documents Invitrogen
LS11636H07E50
50μg Each for $654.00
 
Description

Description

Aldehyde Dehydrogenase 1 family, member A3 (ALDH1A3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
220
PAGE
ALDH1A6, ALDH6, RALDH3

For Research Use Only. Not for use in diagnostic procedures.