missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALDH1A3 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ALDH1A3 (NP_000684.2) (Ala 2-Pro 512) fused to a polyhistide tag at the N-terminus

Manufacturer:  Invitrogen LS11636H07E50

 View more versions of this product

Catalog No. 11636H07E50



Description

Description

Aldehyde Dehydrogenase 1 family, member A3 (ALDH1A3), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Lyophilized
57.0kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
Aldehyde Dehydrogenase 1 family, member A3
His Tag
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
220
PAGE
50μg
N-Terminal
ALDH1A6, ALDH6, RALDH3
ALDH1A3
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.