missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALDH1A1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human ALDH1A1 (NP_000680.2) (Ser 2-Ser 501) fused to a polyhistidine tag at the N-terminus

$457.00 - $457.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias ALDH1A1, RP11-151D14.2, ALDC, ALDH-E1, ALDH1, ALDH11, mgC2318, PUMB1, RALDH1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11388H07E25 View Documents Life Technologies
LS11388H07E25
5 x 5μg Each for $457.00
 
Description

Description

Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
ALDH1A1, RP11-151D14.2, ALDC, ALDH-E1, ALDH1, ALDH11, mgC2318, PUMB1, RALDH1
PAGE
Lyophilized
216

For Research Use Only. Not for use in diagnostic procedures.