missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALCAM (CD166) Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 527) of mouse ALCAM precursor (NP_033785.1) fused to a polyhistidine tag at the C-terminus

$271.00 - $271.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI853494, BEN, CD166, DM-GRASP, mgC27910, MuSC, SC1, ALCAM
Gene ID (Entrez) 11658
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50005M08H25 View Documents Invitrogen
LS50005M08H25
5 x 5μg Each for $271.00
 
Description

Description

Activated leukocyte cell adhesion molecule (ALCAM), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
11658
<1 EU/μg
AI853494, BEN, CD166, DM-GRASP, mgC27910, MuSC, SC1, ALCAM

For Research Use Only. Not for use in diagnostic procedures.