missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

ALCAM (CD166) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) fused to a polyhistidine tag at the C-terminus

$242.00 - $304.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD166, FLJ38514, MEMD, mgC71733, ALCAM
Gene ID (Entrez) 214
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10045H08H25 View Documents Invitrogen
LS10045H08H25
5 x 5μg Each for $242.00
 
10045H08H50 View Documents Invitrogen
LS10045H08H50
50μg Each for $304.00
 
Description

Description

Activated leukocyte cell adhesion molecule (ALCAM), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
214
<1 EU/μg
CD166, FLJ38514, MEMD, mgC71733, ALCAM

For Research Use Only. Not for use in diagnostic procedures.