Please login to your online account to display your discounted pricing

Sino Biological™ ALCAM (CD166) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 526) of human ALCAM (NP_001618.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10045H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10045H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 57.4kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant/Cell Surface Protein
Protein Subtype Glycoproteins
Endotoxin Concentration <1 EU/μg
Gene Alias CD166, FLJ38514, MEMD, mgC71733, ALCAM
Common Name Activated leukocyte cell adhesion molecule
Gene Symbol ALCAM
Protein Tag His Tag
Validated Application Assay Standard
Protein Family CD Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 214

Activated leukocyte cell adhesion molecule (ALCAM), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.

  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.