missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ AKR1B1 Recombinant Human Protein, N-His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human AKR1B1 (P15121) (Met 1-Phe 316) fused to a polyhistidine tag at the N-terminus

$206.00 - $894.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Format Frozen
Gene Alias ADR, ALDR1, ALR2, AR, mgC1804
Gene ID (Entrez) 231
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11294H07E25 Life Technologies
LS11294H07E25
5 x 5μg Each for $206.00
11294H07EL Life Technologies
LS11294H07E250
5 x 50μg Each for $894.00
Specifications

Description

Aldo-Keto Reductase family 1 member B1 (aldose reductase) (AKR1B1), recombinant human protein is supplied frozen. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: PBS, 20% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥85% by SDS-PAGE
Frozen
231
PAGE
ADR, ALDR1, ALR2, AR, mgC1804